Skip to Content
MilliporeSigma
All Photos(1)

Documents

566322-M

Millipore

SIRT1 Inhibitor III

The SIRT1 Inhibitor III, also referenced under CAS 49843-98-3, controls the biological activity of SIRT1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT1 Inhibitor III, 6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide, racemic, SIRT1 Inhibitor III, SIRT2 Inhibitor IV, SIRT3 Inhibitor I, EX-527

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13ClN2O
CAS Number:
Molecular Weight:
248.71
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

storage temp.

2-8°C

InChI

1S/C13H13ClN2O/c14-7-4-5-11-10(6-7)8-2-1-3-9(13(15)17)12(8)16-11/h4-6,9,16H,1-3H2,(H2,15,17)

InChI key

FUZYTVDVLBBXDL-UHFFFAOYSA-N

General description

A cell-permeable indole compound that acts as a potent and highly selective inhibitor of SIRT1 (IC50 = 98 nM). It inhibits other sirtuin family deacetylases only at much higher concentrations (IC50 = 19.6 and 48.7 µM for SIRT2 and SIRT3, respectively) and shows no inhibitory effect against class I and II HDACs or NAD glycohydrolase even at concentrations as high as 100 µM. Shown to be orally bioavailable with a serum half-life of 136 minutes in mice in vivo.

Biochem/physiol Actions

Primary Target
SIRT1
Target IC50: 98 nM against SIRT1

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Solomon, J.M., et al. 2006. Mol. Cell. Biol.26, 28.
Napper, A.D., et al. 2005. J. Med. Chem.48, 8045.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service