Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

566330-M

Millipore

SIRT1/2 Inhibitor VIII, Salermide

The SIRT1/2 Inhibitor VIII, Salermide, also referenced under CAS 1105698-15-4, controls the biological activity of SIRT1/2. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT1/2 Inhibitor VIII, Salermide, N-(3-((2-Hydroxynaphthalen-1-ylmethylene)-amino)-phenyl)-2-phenyl-propionamide, SIRT1 Inhibitor VI, SIRT2 Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22N2O2
CAS Number:
Molecular Weight:
394.47
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

N(c2cc(ccc2)N=Cc3c4c(ccc3O)cccc4)C(=O)C(C)c1ccccc1

InChI

1S/C26H22N2O2/c1-18(19-8-3-2-4-9-19)26(30)28-22-12-7-11-21(16-22)27-17-24-23-13-6-5-10-20(23)14-15-25(24)29/h2-18,29H,1H3,(H,28,30)

InChI key

HQSSEGBEYORUBY-UHFFFAOYSA-N

General description

A cell-permeable 2-hydroxy-naphthaldehyde that acts as an inhibitor against sirtuins SirT1 and SirT2, members of class III HDACs. Salermide effectively inhibits the activity of both SirT1 and SirT2 (by 80% at 100 and 25 µM, respectively), while its structural analogue Sirtinol (Cat. Nos. 566320 and 566321), at 100 µM concentration, inhibits SirT2 only by up to 60% and is of no effect against SirT1. Salermide is also shown to be at least 2-fold more potent than Sirtinol (both at 100 µM) in killing leukemia KG1A and lymphoma Raji cultures.

Warning

Toxicity: Standard Handling (A)

Other Notes

Lara, E., et al. 2009. Oncogene28, 781.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service