Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33169

Sigma-Aldrich

BET Inhibitor, I-BET726

Synonym(s):

BET Inhibitor, I-BET726, BET Inhibitor, BET726, GSK1324726A, 4-((2S,4R)-1-Acetyl-4-((4-chlorophenyl)amino)-2-methyl-1,2,3,4-tetrahydroquinolin-6-yl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23ClN2O3
CAS Number:
Molecular Weight:
434.91
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrahydroquinoline based compound that acts as a highly potent and selective inhibitor of bromodomain and extra-terminal (BET) family proteins (IC50 = 41, 31, and 22 nM for BRD2, BRD3, BRD4, respectively). Binds to the acetyl-lysine recognition pocket of BET proteins and competes with tetra-acetylated histone H4 peptides (K5ac, K8ac, K12ac, K16ac) for binding to the bromodomains of these proteins. Does not affect the activity of over 50 different receptors, ion channels, and transporters even at higher concentrations (1-10 µM). Exhibits cytotoxicity towards neuroblastoma cell lines, induces apoptosis, and inhibits their growth (gIC50 = 26 and 60 nM for CHP-212 and SK-N-AS cells, respectively). Suppresses the growth of human neuroblastoma xenografts in murine models (5 mg/kg, p.o. for 2 weeks) and reduces the expression of MYCN and BCL2. Shown to improve survival rate of Balb/c mice subjected to LPS-induced (15 - 20 mg/kg) septic shock (10 mg/kg, i.v.). Displays desirable pharmacokinetic properties (t1/2 = 3.6 h; AUC 0-t = 6.03 µg/h/ml at 10 mg/kg i.v. in mice).
A cell-permeable tetrahydroquinoline based compound that acts as a highly potent and selective inhibitor of bromodomain and extra-terminal (BET) family proteins (IC50 = 41, 31, and 22 nM for BRD2, BRD3, BRD4, respectively). Binds to the acetyl-lysine recognition pocket of BET proteins and competes with tetra-acetylated histone H4 peptides (K5ac, K8ac, K12ac, K16ac) for binding to the bromodomains of these proteins. Does not affect the activity of over 50 different receptors, ion channels, and transporters even at higher concentrations (1-10 µM). Exhibits cytotoxicity towards neuroblastoma cell lines, induces apoptosis, and inhibits their growth (gIC50 = 26 and 60 nM for CHP-212 and SK-N-AS cells, respectively). Suppresses the growth of human neuroblastoma xenografts in murine models (5 mg/kg, p.o. for 2 weeks) and reduces the expression of MYCN and BCL2. Shown to improve survival rate of Balb/c mice subjected to LPS-induced (15 - 20 mg/kg) septic shock (10 mg/kg, i.v.). Displays desirable pharmacokinetic properties (t1/2 = 3.6 h; AUC 0-t = 6.03 µg/h/ml at 10 mg/kg i.v. in mice).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
BET Inhibitor, I-BET726, is a cell-permeable, potent, and selective inhibitor of BET family proteins (IC₅₀ = 41, 31, and 22 nM for BRD2, BRD3, BRD4, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BET
Target IC50: 41 nM, 31 nM, and 22 nM for BRD2, BRD3, BRD4, BRDT respectively
Target Ki: 4.4 nM for BRD4

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gosmini, R., et al. 2014. J. Med. Chem.57, 811.
Wyce, A., et al. 2013. Plos One.8, e72967.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service