Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0046

Sigma-Aldrich

Azidodifluoroacetamide

Synonym(s):

2-Azido-2,2-difluoroacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$682.00
100 MG
$975.00

$682.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$682.00
100 MG
$975.00

About This Item

Empirical Formula (Hill Notation):
C2H2F2N4O
CAS Number:
Molecular Weight:
136.06
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.06

$682.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

SMILES string

NC(C(N=[N+]=[N-])(F)F)=O

InChI

1S/C2H2F2N4O/c3-2(4,1(5)9)7-8-6/h(H2,5,9)

InChI key

ISQQWTYWSPQUSN-UHFFFAOYSA-N

General description

Azidodifluoroacetamide is a fluoroalkyl azide reagent active in copper-catalyzed azide-alkyne cycloaddition (click reaction). In addition to its use as a highly fluorinated building block, it can also be functionalized onto biomolecules, advanced materials, or polymers containing an alkyne. It is an effective 19F NMR sensor for these materials, particularly in cases where aqueous solubility is necessary and organic solvents cannot be used.

Learn more about fluorinated click chemistry

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorinated azides are a new tool that can be used to introduce fluorinated groups to molecules. These molecules can utilize click chemistry to easily add fluorinated groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service