Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31535

Sigma-Aldrich

AP-1/NF-κB Dual Inhibitor, SP100030

Synonym(s):

AP-1/NF-κB Dual Inhibitor, SP100030, 2-Chloro-4-(trifluoromethyl)pyrimidine-5-N-(3ʹ,5ʹ-bis(trifluoromethyl)phenyl)-carboxamide, N-(3,5-bis(Trifluoromethyl)phenyl)-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxamide, Activator Protein-1 Activation Inhibitor; AP-1 Acti, SP-100030, Nuclear Factor-κB Activation Inhibitor XI, 2-Chloro-4-(trifluoromethyl)pyrimidine-5-N-(3ʹ,5ʹ-bis(trifluoromethyl)phenyl)-carboxamide, N-(3,5-bis(Trifluoromethyl)phenyl)-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxamide, Activator Protein-1 Activation Inhibitor; AP-1 Activation Inhibitor, NF-κB Activation Inhibitor XI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H5ClF9N3O
CAS Number:
Molecular Weight:
437.65
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, conformationally restricted pyrimidinecarboxamide compound that is reported to inhibit against the activation of AP-1- & NF-κB-, but not &beta-actin-, mediated transcriptions (IC50 = 50 nM in Jurkat-based reporter assays; 0.5 h drug pretreatment prior to 5 h stimulation with 1 µg/mL PHA/526511 & 50 ng/mL PMA/500582 & 524400) and target genes expressions, including IL-2, IL-8, TNF-α, in 6 T cell cultures (by >85% at 3 µM), while exhibiting little inhibitory potency against cytokine inductions in 10 non-T cell cultures (by <5% at 10 µM), including endothelial, epithelial, fibroblast, monocytic, and osteoblast cultures. Intraperitoneal injection is shown to display in vivo anti-inflammatory efficacy in a murine model of collagen-induced arthritis (10 mg/kg/d) and a rodent model of Ova-induced asthma response (20 mg/kg/d). Dual AP-1 & NF-κB inhibition via daily SP100030 subcutaneous injection is also reported to suppress AH-130 Yoshida ascites hepatoma tumor expansion in rats in vivo (5 mg/kg/d).
A cell-permeable, non-toxic, conformationally restricted pyrimidinecarboxamide compound that acts as a potent, reversible T-cell specific inhibitor of both AP-1 and NF-κB transcriptional activation (IC50 = 50 nM). Reduces DNA binding activity of NF-κB and downregulates NF-κB driven cytokine gene expression. Displays only trivial inhibitory effect against cytokine induction in a variety of other cells. Exhibits anti-inflammatory activity in a murine model of collagen-induced arthritis (~ 10 mg/kg/d, i.p.) and suppresses the expansion of AH-130 Yoshida ascites hepatoma in rats (~ 5 mg/kg/d, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
AP-1 and NF-κB
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Ye, N., et al. 2014. J. Med. Chem.57, 6930.
Moore-Carrasco, R., et al. 2009. Anticancer Res.29, 1315.

Huang, T.J., et al. 2001. Br. J. Pharmacol.134, 1029.
Gerlag, D.M., et al. 2000. J. Immunol.165,1652.
Sullivan, R.W., et al. 1998. J. Med. Chem.41, 413.
Goldman, M.E., et al. 1996. Transplant. Proc.28, 3106.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service