Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TMS-011

Sigma-Aldrich

Endotoxin-Free Ultra Pure Water

Cell Culture

Synonym(s):

H20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.71
technique(s):
cell culture | mammalian: suitable

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

General description

Endotoxin-Free Ultra Pure Water is sterile filtered through a 0.1 micron filter and is considered endotoxin-free (<0.005 EU/ml)

Application

Endotoxin-free ultra pure water has been used in the purification of cationic lipid nanoparticles. It has also been used to purify graphite particles.

Quality

Appearance: Clear Liquid/No Particulates
Sterility Testing: Negative for Growth
pH: 5.0-7.0
Endotoxin: <0.005 EU/mL

Storage and Stability

Store at room temperature until expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fatima Alnasser et al.
Nano letters, 19(2), 1260-1268 (2019-01-11)
The biological interactions of graphene have been extensively investigated over the last 10 years. However, very little is known about graphene interactions with the cell surface and how the graphene internalization process is driven and mediated by specific recognition sites
Daniel Helman et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 85(2), 162-168 (2013-06-15)
We developed a membrane bound reporter and selection molecule for sorting by fluorescence activated cell sorting (FACS) of cells producing a protein of interest. This molecule is composed of a transmembrane (TM) domain, fused on its extracellular end to a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service