Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30821

Sigma-Aldrich

PTP1B Inhibitor II, JTT-551

Synonym(s):

PTP1B Inhibitor II, JTT-551, 2-(((5-tert-Butylthiazol-2-yl)methyl)((4-(4-((4-heptan-4-yl)phenoxy)methyl)phenyl)thiazol-2-yl)methyl)amino)acetic acid, 2HCl, Protein Tyrosine Phosphatase 1B Inhibitor II; PTP Inhibitor XVI, PTPN2/TCPTP Inhibitor IV, (((5-(1,1-Dimethylethyl)thiazol-2-yl)methyl)(((4-(4-(4-(1-propylbutyl)phenoxy)methyl)phenl)thiazol-2-yl)methyl)amino)acetic acid, 2HCl, JTT551

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H43N3O3S2 · 2HCl
Molecular Weight:
678.78
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

CCCC(CCC)C1=CC=C(C=C1)OCC2=CC=C(C=C2)C3=CSC(=N3)CN(CC4=NC=C(S4)C(C)(C)C)CC(=O)O

General description

A cell-permeable dithiazolylmethylglycinate that acts as a PTP1B-selective tyrosine phosphatase inhibitor (Ki = 220 nM/PTP1B, 9.3 µM/TCPTP, >30 µM/CD45, >30 µM/LAR; Substrate = pNPP, Cat. nos. 6535-OP & 6537-OP) with a mixed-type mode of inhibition. Shown to enhance insulin-stimulated glucose uptake by rat L6 myotubes (by 17% and 36% in 10 min, respectively, at 10 and 30 µM; drug treatment starts 2 h prior to 10 nM insulin stimulation) in vitro and significantly enhance insulin-induced IR tyrosine phosphorylation (1 min) in liver of overnight fasted ob/ob mice (59% vs. 39% of C57BL/6J mice level, respectively, with or without single 10 mg/kg oral dosage 3 h prior to 3 U/kg insulin i.v. injection) in vivo. Daily oral administration is efficacious in reducing blood glucose and triglyceride levels among ob/ob (10 mg/kg/d) and db/db mice (30 mg/kg/d) without significant effect on body weight, while blood insulin level reduction is also seen among ob/ob, but not db/db mice.
A cell-permeable, orally available dithiazolylmethylglycinate that acts as a potent, selective, and reversible inhibitor of protein tyrosine phosphatase 1B (PTP1B; Ki = 220 nM). The inhibition appears to be of mixed type. Does not affect the activities of TCPTP, CD45, and LAR phosphatases (Ki = 9.3, >30 and >30 µM, respectively). Shown to increase insulin-stimulated glucose uptake in L6 rat skeletal muscle cells (~10-30 µM). Enhances insulin receptor phosphorylation in liver of ob/ob mice and reduces blood glucose level in a dose-dependent manner with hypoglycemic effects evident at >1 mg/kg (p.o.). Also effective in reducing blood glucose levels in db/db mice (>3 mg/kg, p.o.) without causing any weight gain.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PTP1B
Reversible: yes
Target Ki: 220 nM for PTP1B

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a diHCl salt.

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Fukuda, S., et al. 2010. Diabetes Obes. Metab.12, 299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service