Skip to Content
MilliporeSigma
All Photos(1)

Documents

539749

Sigma-Aldrich

PTP1B Inhibitor

InSolution, ≥97%

Synonym(s):

InSolution PTP1B Inhibitor, PTP Inhibitor XXII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H19Br2N3O7S3
Molecular Weight:
741.45
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, selective, reversible, and non-competitive allosteric inhibitor of PTP1B (IC50 = 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively). Binds to a novel site away from the catalytic pocket and inhibits PTP1B activity by preventing closure of the WPD loop.

Biochem/physiol Actions

Cell permeable: yes
IC50 = 4 and 8 µM for PTP1B403 and PTP1B298, respectively
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (2 mg/270 µl) solution of PTP1B Inhibitor (Cat. No. 539741) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Wiesmann, C., et al. 2004. Nat. Struct. Mol. Biol.11, 730.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service