Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528240

Sigma-Aldrich

Platelet Membrane Glycoproteins, IIbIIIa, Human Platelets

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$885.00

$885.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$885.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$885.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

General description

Native, platelet membrane glycoprotein IIbIIIaA from human platelets. A platelet integrin that binds to fibrinogen via the recognition sequence Arg-Gly-Asp-Ser. Acts as a receptor for such adhesive ligands as fibrinogen, fibronectin and von Willebrand factor during platelet stimulation. Glycoproteins IIb and IIIa constitute the fibrinogen receptor and are required for platelet aggregation. Glycoprotein IIb consists of two disulfide-linked subunits, GPIIbα and GPIIbβ, whereas GPIIIa has only one polypeptide chain.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, 1 mM CaCl2, 50% glycerol, ≤0.1% sodium azide, pH 7.4.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Avoid freeze/thaw cycles.

Other Notes

Schwarz M., et al. 2004. J. Pharmacol. Exp. Therap.308, 1002.
Wippler, J., et al. 1994. J. Biol. Chem. 269, 8754.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service