Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT893M

Sigma-Aldrich

Anti-Platelet Antibody, clone PAC-1

clone PAC-1, from mouse

Synonym(s):

Integrin alpha-IIb, GPalpha IIb, GPIIb, Platelet membrane glycoprotein IIb, CD41)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PAC-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
dot blot: suitable
flow cytometry: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ITGA2B(3674)

General description

Integrin alpha-IIb (UniProt: P08514; also known as GPalpha IIb, GPIIb, Platelet membrane glycoprotein IIb, CD41) is encoded by the ITGA2B (also known as GP2B, ITGAB) gene (Gene ID: 3674) in human. GPIIb is a single-pass type I membrane protein of the integrin alpha chain family. It serves as a receptor of fibronectin, fibrinogen, plasminogen, prothrombin, thrombospondin and vitronectin. It recognizes the sequence R-G-D in a wide array of ligands. . Following activation it brings about platelet/platelet interaction through binding of soluble fibrinogen. This step leads to rapid platelet aggregation, which physically plugs ruptured endothelial cell surface. GPIIb is synthesized with a signal peptide of 31 amino acids that is cleaved to produce the mature form that has an extracellular domain (aa 32-993), a helical domain (aa 994-1019), and a cytoplasmic tail (aa 1020-1039). Three isoforms of GPII b are reported that are generated by alternative splicing. Isoform 1 and 2 have been identified in platelets and megakaryocytes, but not in reticulocytes or in Jurkat and U-937 white blood cell line. Isoform 3 is expressed by leukemia, prostate adenocarcinoma and melanoma cells but not by platelets or normal prostate or breast epithelial cells. Clone PAC- 1 is a pentameric IgM that is reported to bind to the activated platelets, but not to unstimulated platelets.

Specificity

Clone PAC-1 is a pentameric IgM that binds to the activated platelets, but not to unstimulated platelets.

Immunogen

Platelets obtained from the blood of a patient with Glanzmann′s thrombasthenia.

Application

Anti-Platelet, clone PAC-1, Cat. No. MABT893 is a mouse monoclonal antibody that detects Integrin alpha-IIb and has been tested for use in Dot Blot, ELISA, Flow Cytometry, Radioimmunoassay, and Western Blotting.
Dot Blot Analysis: A representative lot detected Platelet in Dot Blot applications (Shattil, S.J., et. al. (1985). J Biol Chem. 260(20):11107-14).

ELISA Analysis: A representative lot detected Platelet in ELISA applications (Shattil, S.J., et. al. (1985). J Biol Chem. 260(20):11107-14).

Radioimmunoassay Analysis: A representative lot detected Platelet in Radioimmunoassy applications (Shattil, S.J., et. al. (1985). J Biol Chem. 260(20):11107-14; Shattil, S.J., et. al. (1987). Blood. 70(1):307-15).

Western Blotting Analysis: A representative lot detected Platelet in Western Blotting applications (Shattil, S.J., et. al. (1985). J Biol Chem. 260(20):11107-14).

Flow Cytometry Analysis: A representative lot detected Platelet in Flow Cytometry applications (Caron, A., et. al. (2002). J Cardiovasc Pharmacol. 40(2):296-306; Taub, R., et. al. (1989). J Biol Chem. 264(1):259-65; Shattil, S.J., et. al. (1987). Blood. 70(1):307-15).

Flow Cytometry Analysis: A 1:200 dilution from a representative lot detected Platelet in freshly isolated human platelets activated with convulxin (Courtesy of Shuchi Gupta, PhD, University of Pennsylvania, USA).
Research Category
Cell Structure

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgM.

Target description

113.38 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service