Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

527962

Millipore

Pimaricin, Streptomyces chattanoogensis

Synonym(s):

Pimaricin, Streptomyces chattanoogensis, Natamycin, Pimafucin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H47NO13
CAS Number:
Molecular Weight:
665.73
MDL number:
UNSPSC Code:
12352200

Assay

≥80% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

acetic acid: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

General description

Antifungal polyene macrolide that is active in the cell membrane.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Pederson, J.C. 1992. Appl. Environ. Microbiol. 58, 1064.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service