Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521225

Sigma-Aldrich

PDGF, BB Homodimer, Human, Recombinant, E. coli

Synonym(s):

rhPDGF-BB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 ng/μg Endotoxin (ng/μg PDGF-BB)

shipped in

dry ice

storage temp.

−70°C

General description

Recombinant, human platelet-derived growth factor (disulfide-linked dimer of two 109 amino acid B-chain monomers) expressed in E. coli. Shown to be a potent chemoattractant for neutrophils, mesenchymal and mononuclear cells. Has the same amino acid sequence as mature human PDGF-B. Rapidly activates protein kinase D in vascular smooth muscle cells.
Recombinant, human platelet-derived growth factor (disulfide-linked dimer of two 109 amino acid B-chain monomers) expressed in E. coli. Shown to be a potent chemoattractant for neutrophils, mesenchymal, and mononuclear cells. PDGF represents one of the major mitogens present in platelets. It exerts its effects by binding to specific high-affinity dimeric receptor tyrosine kinases. Material has the same amino acid sequence as mature human PDGF-B. Two distinct PDGF receptors have been identified: the PDGFα receptor, which binds to all three forms of PDGF, and the PDGFβ receptor, which can bind only PDGF-BB and PDGF-AB.

Biochem/physiol Actions

ED₅₀ = 1-5 ng/ml as determined by the dose dependent proliferation of BALB/c 3T3 cells

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a sterile-filtered solution, carrier free.

Reconstitution

Reconstitute to a concentration of ≥100 µg/ml 100 mM acetic acid, 0.1% BSA. Further dilute with low endotoxin medium or buffered solution containing FBS or tissue culture grade BSA just prior to use.

Other Notes

Abedi, H., et al. 1998. FEBS Lett. 427, 209.
Abboud, H.E., et al. 1994. J. Cell Physiol. 158, 140.
Westermark, B., and Heldin, C.-H. 1991. Cancer Res.51, 5087.
Raines, E.W., et al. 1985. Methods Enzymol.109, 749.
Johnson, A., et al. 1984. EMBO J.3, 921.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulina Villar-Fincheira et al.
Frontiers in physiology, 12, 722528-722528 (2021-10-29)
Little is known about the effects of training load on exercise-induced plasma increase of interleukin-6 (IL-6) and soluble IL-6 receptor (sIL-6R) and their relationship with vascular remodeling. We sought to evaluate the role of sIL 6R as a regulator of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service