Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

517312

Sigma-Aldrich

Anti-P-Glycoprotein Mouse mAb (C494)

liquid, clone C494, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C494, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, hamster

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Purified mouse monoclonal antibody. Recognizes the ~170 kDa MDR1 isoform of P-glycoprotein.
Recognizes the ~170 kDa MDR1 isoform of P-glycoprotein. Does not cross-react with the MDR3 P-glycoprotein.
This Anti-P-Glycoprotein Mouse mAb (C494) is validated for use in Paraffin Sections, FC, Frozen Sections, Immunoblotting, ICC, IP for the detection of P-Glycoprotein.

Immunogen

Epitope: gene-specific internal cellular epitope present only on the MDR1 isoform of P-glycoprotein
Hamster and Human
SDS-solubilized plasma membranes of a multidrug resistant (MDR) Chinese hamster ovary (CHO) cell line and a human leukemic cell line

Application

Paraffin Sections (5-10 µg/ml, see comments)

Flow Cytometry (see comments)

Frozen Sections (5-10 µg/ml, see comments)

Immunoblotting (see comments)

Immunocytochemistry (see application references)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 1% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Cell lines or frozen tissue, such as human liver or colon.

Other Notes

Does not cross-react with MDR3 P-glycoprotein. Also reported to work for flow cytometry, immunoblotting, and immunoprecipitation. For immunocytochemistry, frozen or paraffin-embedded material may be used. Permeabilization of the cell membrane is required to expose the epitope recognized by this antibody. Variables associated with assay conditions will dictate the proper working dilutions.
Rao, V.V., et al. 1994. Cancer Res.54, 1536.
Toth, K., et al. 1992. Am. J. Pathol.140, 1009.
Georges, E., et al. 1990. Proc. Natl. Acad. Sci. USA87, 152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regino Mercado-Lubo et al.
Nature communications, 7, 12225-12225 (2016-07-28)
Salmonella enterica serotype Typhimurium is a food-borne pathogen that also selectively grows in tumours and functionally decreases P-glycoprotein (P-gp), a multidrug resistance transporter. Here we report that the Salmonella type III secretion effector, SipA, is responsible for P-gp modulation through
Amélie Petitprez et al.
International journal of oncology, 42(5), 1644-1653 (2013-04-03)
Irinotecan is a major anticancer agent specifically targeting DNA topoisomerase I. Its cytotoxicity is mediated via a two-step process involving accumulation of reversible DNA‑topoisomerase I complexes associated with transient DNA single-strand breaks which subsequently are converted into permanent DNA double-strand
Golam Kibria et al.
Biological & pharmaceutical bulletin, 37(12), 1926-1935 (2014-12-03)
Multi-drug resistance (MDR) of cancers to chemotherapy including doxorubicin (DOX) is mediated by several factors. To design an effective therapy for the treatment of chemotherapy-resistant cancers, it is essential to explore the elements responsible for mediating MDR. However, exploring these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service