Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09226

Sigma-Aldrich

PI 3-K Inhibitor XXI, GDC-0941

Synonym(s):

PI 3-K Inhibitor XXI, GDC-0941, GDC0941, 2-(1H-Indazol-4-yl)-6-(4-methylsulfonyl-piperazin-1-ylmethyl)-4-morpholin-4-yl-thieno[3,2- d]pyrimidine, 4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulfonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$247.00

$247.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C23H27N7O3S2
CAS Number:
Molecular Weight:
513.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$247.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N1CCN(CC1)Cc2[s]c3c(nc(nc3N6CCOCC6)c4c5c(n[nH]c5)ccc4)c2)C

InChI

1S/C23H27N7O3S2/c1-35(31,32)30-7-5-28(6-8-30)15-16-13-20-21(34-16)23(29-9-11-33-12-10-29)26-22(25-20)17-3-2-4-19-18(17)14-24-27-19/h2-4,13-14H,5-12,15H2,1H3,(H,24,27)

InChI key

LHNIIDJUOCFXAP-UHFFFAOYSA-N

General description

A cell-permeable thienopyrimidine compound that acts as a potent, ATP-competitive inhibitor against class IA (IC50 = 3 nM/p110α, 33 nM/p110β, 3 nM/p110δ) & IB (IC50 = 75 nM/p110γ) PI 3-K kinases, including the two known p110α hot spot mutants E454K & H1047R (IC50 = 3 nM), while exhibiting much reduced or little potency against more than 200 other kinases, including TrkA, class II C2β, class IV mTOR & DNA-PK (IC50 = 2.85, 0.67, 0.58, and 1.23 µM, respectively) and class III Vps34 (IC50 >10 µM). Effectively inhibits cellular Akt S473 phosphorylation (IC50 = 46 nM/U87MG, 37 nM/PC3, and 28 nM/MDA-MB-361) as well as proliferation in cancer cultures in vitro (GI50 = 950/U87MG, 280/PC3, and 720 nM/MDA-MB-361) and greatly prevents the expansion of U87MG glioblastoma xenograft in mice in vivo (by 83%; 75 mg/kg/d p.o. for 21 days). Oral bioavailability is reported in human, dog, mouse, and rat.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI 3-K
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Tabe, Y., et al. 2014. Acta Haematol.131, 59.
Ware, J.A., et al. 2013. Mol. Pharm.10, 4047.
Junttila,.T., et al. 2009. Cancer Cell15, 429.
Raynaud, F.I., et al. 2009. Mol Cancer Ther.8, 1725.
Folkes, A. J., et al. 2008. J. Med. Chem.51, 5522.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service