Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09665

Sigma-Aldrich

Glucokinase Activator III, Ro-28-1675

Synonym(s):

Glucokinase Activator III, Ro-28-1675, GK Activator III, Hexokinase D Activator III, Hexokinase IV Activator III, Ro-28-0450 ( R)-enantiomer, RO0281675, GK Activator III, Hexokinase D Activator III, Hexokinase IV Activator III, Ro-28-0450 (R)-enantiomer, RO0281675

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$283.00

$283.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$283.00

About This Item

Empirical Formula (Hill Notation):
C18H22N2O3S2
CAS Number:
Molecular Weight:
378.51
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$283.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(C)c1ccc(cc1)[C@@H](CC3CCCC3)C(=O)Nc2[s]ccn2

InChI

1S/C18H22N2O3S2/c1-25(22,23)15-8-6-14(7-9-15)16(12-13-4-2-3-5-13)17(21)20-18-19-10-11-24-18/h6-11,13,16H,2-5,12H2,1H3,(H,19,20,21)/t16-/m1/s1

InChI key

NEQSWPCDHDQINX-MRXNPFEDSA-N

General description

A cell-permeable thiazolyl-propanamide compound that acts as an allosteric glucokinase-(GK) selective activator, increasing Vmax and decreasing [S]1/2 (by 1.5- and 4.3-fold, respectively, at 3 µM; [Glucose] = 5 mM) in human GK enzymatic assays, as well as freeing GK from the inhibitory action of GK regulatory protein (GKRP), resulting in increased GK nuclear-to-cytosol translocation in primary rat hepatocytes (EC50/[Glucose] = 0.16 µM/16.8 mM, 0.27 µM/8.4 mM, 2.2 µM/2.8 mM) and reduced glucose concentration for stimulating insulin secretion from isolated rat pancreatic islets (threshold [glucose] = 3 mM vs 6 mM with or without 3 µM Ro-28-1675). Exhibits in vivo glucose-lowering and insulin-releasing activity in non-diabetic C57BL/6 mice and Wistar rats and is efficacious in several murine (KK/Upj-Ay/J, ob/ob, and diet-induced obese C57BL/6) and rodent (Goto-Kakizaki) type II diabetes mellitus (T2DM) models (10 to 50 mg/kg via p.o.), while efficacy is no longer observed in older hypoinsulinemic db/db mice with blood glucose levels reaching 300 mg/dl. Reported not to affect the activity of hexokinase I or II and display no activity toward GK-independent insulin release from β cells upon 2-Ketoisocaproic acid (α-KIC) stimulation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Glucokinase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wolff, M., et al. 2008. J. Biomol. Screen13, 837.

Grimsby, J., et al. 2003. Science301, 370.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service