Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346021

Sigma-Aldrich

Glucokinase Activator, Cpd A

The Glucokinase Activator, Cpd A, also referenced under CAS 603108-44-7, modulates Glucokinase. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Glucokinase Activator, Cpd A, Hexokinase D Activator, Hexokinase IV Activator, 2-Amino-5-(4-methyl-4H-(1,2,4)-triazole-3-yl-sulfanyl)-N-(4-methyl-thiazole-2-yl)benzamide, Compound A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$233.00

$233.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$233.00

About This Item

Empirical Formula (Hill Notation):
C14H14N6OS2
CAS Number:
Molecular Weight:
346.43
MDL number:
UNSPSC Code:
41121706
NACRES:
NA.77

$233.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thiazolylamide glucokinase (GK; Hexokinase IV/D) activator that reversibly targets a allosteric site that is only exposed for binding in the glucose-bound GK conformation, but not in the unbound "super-open" GK conformation or the GKRP- (GK Regulatory Protein) bound GK, and effectively stabilizes GK in an active conformation as well as prevents its interaction with and nuclear sequestration by GKRP, while exhibiting little effect against Hexokinase I/II/III. Compound A is shown to activate GK activity in cell-free assays (EC50 = 0.42 and 0.14 µM in the presence of 2.5 mM and 10 mM of glucose, respectively) and effectively induce cellular GK activation and increase glucose metabolism both in in rats (10 to 30 mg/kg, p.o.) in vivo and in rat islets and hepatocytes cultures (10 to 30 µM) in vitro.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Iino, T., et al. 2009. Bioorg. Med. Chem. Lett.19, 5531.
Mitsuya, M., et al. 2009. Bioorg. Med. Chem. Lett.19, 2718.
Nishimura, T., et al. 2009. Bioorg. Med. Chem. Lett.19, In press.
Futamura, M., et al. 2006. J. Biol. Chem.281, 1357.
Kamata, K., et al. 2004. Structure12, 429.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie L Lewandowski et al.
Cell metabolism, 32(5), 736-750 (2020-11-05)
Pancreatic β cells couple nutrient metabolism with appropriate insulin secretion. Here, we show that pyruvate kinase (PK), which converts ADP and phosphoenolpyruvate (PEP) into ATP and pyruvate, underlies β cell sensing of both glycolytic and mitochondrial fuels. Plasma membrane-localized PK
Ryota Inoue et al.
iScience, 25(7), 104603-104603 (2022-07-09)
Uncoupling protein 2 (UCP2), a mitochondrial protein, is known to be upregulated in pancreatic islets of patients with type 2 diabetes (T2DM); however, the pathological significance of this increase in UCP2 expression is unclear. In this study, we highlight the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service