Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.05880

Sigma-Aldrich

L-745,870 Trihydrochloride

Synonym(s):

L-745,870 Trihydrochloride, Dopamine D₄ receptor Antagonist, L-745,870 trihydrochloride, L-745870, L745870

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4 · 3HCl
CAS Number:
Molecular Weight:
436.21
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.4 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: soluble

storage temp.

2-8°C

InChI

1S/C18H19ClN4/c19-15-3-5-16(6-4-15)23-10-8-22(9-11-23)13-14-12-21-18-17(14)2-1-7-20-18/h1-7,12H,8-11,13H2,(H,20,21)

InChI key

OGJGQVFWEPNYSB-UHFFFAOYSA-N

General description

Selective D4 receptor antagonist with excellent brain penetration. Displays an affinity (Ki) of 0.4 nM for human D4 receptors with >1000-fold binding selectivity over D2 and D3 receptors. Does not block agonist-induced activation of GIRK currents. It is used frequently in retinal and CNS studies of D4 modulation.

Biochem/physiol Actions

Primary Target
D₄
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Li, H. et al., 2013. J. Neurosci.33, 3135.

Pillai, G. et al. 1998. Neuropharmacol.337, 983.
Kulagowski, J. et al. 1996. J. Med. Chem.39, 1941.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service