Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05877

Sigma-Aldrich

(S)-(-)-Raclopride

Synonym(s):

(S)-(-)-Raclopride, Dopamine Receptor D2 Antagonist, (S)-Raclopride, Raclopride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$123.00

$123.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$123.00

About This Item

Empirical Formula (Hill Notation):
C15H20Cl2N2O3
CAS Number:
Molecular Weight:
347.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$123.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mM
ethanol: 25 mM

storage temp.

2-8°C

SMILES string

Clc1c(c(c(c(c1)Cl)O)C(=O)NC[C@@H]2N(CCC2)CC)OC

InChI

1S/C15H20Cl2N2O3/c1-3-19-6-4-5-9(19)8-18-15(21)12-13(20)10(16)7-11(17)14(12)22-2/h7,9,20H,3-6,8H2,1-2H3,(H,18,21)/t9-/m1/s1

InChI key

WAOQONBSWFLFPE-SECBINFHSA-N

General description

D2-like dopamine receptor antagonist targeting D2R (pKi =8.0), D3R (pKi = 7.9), and 5-HT1A R (pKi=5.2). Used to explore D2 like receptor mediated effects in neuropathic pain, and in antagonism studies of DA receptors during heroin usage and cocaine. D2 Receptor selectivity in various preps shows IC50 values around 32 nM.

Biochem/physiol Actions

Reversible: yes

Warning

Toxicity: Standard Handling (A)

Other Notes

Dang, Y. H. et al. 2010. Neurosci.169, 1872.

Platt, D. M. et al. 2010. J. Pharmacol. Exp. Ther.332, 985.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service