Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.00495

Sigma-Aldrich

RubR1 Activator, GF-15

Synonym(s):

RubR1 Activator, GF-15, Supernumerary Centrosomes Clustering Inhibitor, 2ʹ-Benzyloxy-2ʹ-demethoxygriseofulvin, (2 S,6ʹ R)-(7-Chloro-4,6-dimethoxy-benzofuran-3-on)-2-spiro-1ʹ-(2ʹ-benzyloxy-6ʹ-methylcyclohex-2ʹ-en-4ʹ-one), (2, Supernumerary Centrosomes Clustering Inhibitor, 2ʹ-Benzyloxy-2ʹ-demethoxygriseofulvin, (2S,6ʹR)-(7-Chloro-4,6-dimethoxy-benzofuran-3-on)-2-spiro-1ʹ-(2ʹ-benzyloxy-6ʹ-methylcyclohex-2ʹ-en-4ʹ-one), (2S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21ClO6
CAS Number:
Molecular Weight:
428.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 2′-benzyloxy griseofulvin derivative that displays enhanced mammalian cancer-selective toxicity (IC50 ≤2, ≤4, ≤12, and ≥30 µM against panels of leukemia, myeloma, solid tumors, and non-malignant cell lines, respectively), but little antifungal activity, by activating spindle-assembly-checkpoint (Pos. vs. Neg. RubR1 staining of metaphase PC-3 kinetophores with or without 24 h 0.75 µM GF-15 treatment) without affecting microtubule kinetochore attachment (Neg. Mad2 staining of metaphase PC-3 kinetophores with or without GF-15). Cancer cells are reported to undergo multipolar (MP) mitosis with normal or supernumerary centrosomes (Aberrant vs. declustered, respectively) in the presence of GF-15 (%Declustered / %Aberrant MP mitotic cells among all anaphase cells = 5.6/&lt0.5, 10/3.9, 16/18, 28/30, respectively, in PC-3 cultures treated with 0, 0.375, 0.75, or 1.5 µM GF-15 for 24 h), resulting in subsequent apoptotic cell death. Shown to retard OPM2 myeloma- and HT29 colon cancer-derived tumor expansion in mice (by 49% and 56%, respectively at the end of 2-wk treatment; 10 i.p. dosages at 20 mg/kg/day in 2 weeks) in vivo. Based on stability data obtained with its p-iodobenzyl analogue, GF-15 half-life is expected to be least 48 h in cultures.
A cell-permeable 2′-benzyloxy griseofulvin derivative that displays enhanced mammalian cancer-selective toxicity (IC50 ≤2, ≤4, ≤12, and ≥30 µM against panels of leukemia, myeloma, solid tumors, and non-malignant cell lines, respectively), but little antifungal activity, by activating spindle-assembly-checkpoint without affecting microtubule kinetochore attachment. Cancer cells are reported to undergo multipolar (MP) mitosis with normal or supernumerary centrosomes in the presence of GF-15, resulting in subsequent apoptotic cell death. Shown to retard HT29 colon cancer & OPM2 myeloma tumor expansion in mice (20 mg/kg/day i.p.) in vivo. GF-15 half-life is expected to be least 48 h in cultures.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Raab, M.S., et al. 2012. Cancer Res.72, 5374.
Ronnest, M.H., et al. 2009. J. Med. Chem.52, 3342.
Crosse, R., et al. 1964. J. Gen. Microbiol.34, 51.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service