444190
MAP Kinase Inhibitor Set II
The MAP Kinase Inhibitor Set II controls the biological activity of MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Select a Size
$469.00
Select a Size
About This Item
$469.00
Recommended Products
Quality Level
form
solid
potency
2 μM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
shipped in
ambient
storage temp.
−20°C
General description
The Calbiochem MAP Kinase Inhibitor Set II contains 4 inhibitors with different MAP Kinase specificities for your convenience.
Packaging
Warning
Reconstitution
(-20°C).
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service