Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416200

Sigma-Aldrich

Dipeptidylpeptidase IV Inhibitor I

The Dipeptidylpeptidase IV Inhibitor I controls the biological activity of Dipeptidylpeptidase IV. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Dipeptidylpeptidase IV Inhibitor I, Diprotin A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$255.00

$255.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$255.00

About This Item

Empirical Formula (Hill Notation):
C17H31N3O4
Molecular Weight:
341.45
UNSPSC Code:
12352200
NACRES:
NA.77

$255.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

amorphous solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A dipeptidylpeptidase IV (DPP IV) inhibitor that inhibits both endo- and exopeptidase activities of DPPIV.

Biochem/physiol Actions

Cell permeable: no
Primary Target
dipeptidylpeptidase IV (DPP IV)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ile-Pro-Ile

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyunyee Yoon et al.
Biomolecules & therapeutics, 29(2), 154-165 (2020-11-06)
This study aimed to investigate whether the antidiabetic drugs dipeptidyl peptidase 4 (DPP4) inhibitors such as evogliptin and sitagliptin affect the membrane DPP4 (mDPP4) enzymatic activity and immune function of T helper1 (Th1) cells in terms of cytokine expression and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service