Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420323

Sigma-Aldrich

KT5720

InSolution, ≥95%

Synonym(s):

InSolution KT5720, PKA Inhibitor VII

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$181.00

$181.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
$181.00

About This Item

Empirical Formula (Hill Notation):
C32H31N3O5
CAS Number:
Molecular Weight:
537.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$181.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

[n]21c3c4[n](c6c(c4c7c(c3c8c2cccc8)CNC7=O)cccc6)[C@H]5O[C@@]1([C@](C5)(O)C(=O)OCCCCCC)C

InChI

1S/C32H31N3O5/c1-3-4-5-10-15-39-30(37)32(38)16-23-34-21-13-8-6-11-18(21)25-26-20(17-33-29(26)36)24-19-12-7-9-14-22(19)35(28(24)27(25)34)31(32,2)40-23/h6-9,11-14,23,38H,3-5,10,15-17H2,1-2H3,(H,33,36)/t23-,31+,32+/m0/s1

InChI key

ZHEHVZXPFVXKEY-RUAOOFDTSA-N

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target Ki: 56 nM against protein kinase A

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 2 mM (50 µg/47 µl) solution of KT 5720 (Cat. No. 420320) in DMSO.

Reconstitution

Following the initial thaw, aliquot and freeze (20°C).

Other Notes

Simpson, C.S., and Morris, B.J. 1995. Neurosci.68, 97.
Cabell, L, and Audesirk, G. 1993. Int. J. Dev. Neurosci.11, 357.
Gadbois, D.M., et al. 1992. Proc. Natl. Acad. Sci. USA89, 8626.
Kase, H., et al. 1987. Biochem. Biophys. Res. Commun.142, 436.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilse Delint-Ramirez et al.
Molecular cell, 82(20), 3794-3809 (2022-10-08)
Neuronal activity induces topoisomerase IIβ (Top2B) to generate DNA double-strand breaks (DSBs) within the promoters of neuronal early response genes (ERGs) and facilitate their transcription, and yet, the mechanisms that control Top2B-mediated DSB formation are unknown. Here, we report that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service