Skip to Content
MilliporeSigma
All Photos(1)

Documents

420107-M

Millipore

Jasplakinolide, Jaspis johnstoni

A cell-permeable F-actin probe.

Synonym(s):

Jasplakinolide, Jaspis johnstoni - CAS 102396-24-7 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H45BrN4O6
CAS Number:
Molecular Weight:
709.67
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

InChI

1S/C36H45BrN4O6/c1-20-15-21(2)17-23(4)47-32(43)19-30(25-11-13-26(42)14-12-25)40-35(45)31(18-28-27-9-7-8-10-29(27)39-33(28)37)41(6)36(46)24(5)38-34(44)22(3)16-20/h7-15,21-24,30-31,39,42H,16-19H2,1-6H3,(H,38,44)(H,40,45)/b20-15+/t21-,22+,23-,24-,30+,31+/m0/s1

InChI key

GQWYWHOHRVVHAP-HZIRKPFGSA-N

General description

A cell-permeable F-actin probe. A cyclodepsipeptide with antifungal and antitumor properties. A potent inducer of actin polymerization and stabilization in vitro. Prevents activation of store-mediated Ca2+ entry into cells. It competes with Phalloidin (Cat. No. 516640) for actin binding (Kd = 15 nM) and enhances apoptosis induced by interleukin 2 (IL-2) deprivation. A 1 mM (50 µg/71 µl) solution of Jasplakinolide, Jaspis johnstoni (Cat. No. 420127) in DMSO is also available.

Biochem/physiol Actions

Kd = 15 nM for actin binding
Primary Target
Inducer of actin polymerization and stabilization in vitro

Warning

Toxicity: Harmful (C)

Other Notes

Rosado, J.A., et al. 2000. J. Biol. Chem. 275, 7527.
Posey, S.C., and Bierer, B.E. 1999. J. Biol. Chem. 274, 4259.
Senderowicz, A.M., et al. 1995. J. Natl. Cancer Inst. 87, 46.
Bubb, M.R., et al. 1994. J. Biol. Chem.269, 14869.
Scott, V.R., et al. 1988. Antimicrob. Agents Chemother.32, 1154.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service