Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

401455-M

Millipore

Goat Anti-Human IgG, H&L Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase (HRP). Recognizes human IgG, heavy and light chains.
This Goat Anti-Human IgG, H&L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Human IgG, H&L Chain Specific.

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

0.01M PBS, pH 7.6, 10% mannose, 2% sucrose, 0.5 mM EDTA, 1% BSA, 0.1 % Kathon

Other Notes

Monospecific for human IgG, heavy and light chains, as determined by immunoelectrophoresis against normal human serum. o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used for testing immunoenzyme reactivity. Variables associated with assay conditions will dictate the proper working dilutions.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Yang et al.
Cellular and molecular biology (Noisy-le-Grand, France), 69(10), 166-173 (2023-11-13)
Cervical squamous cell carcinoma and endocervical adenocarcinoma (CESC) is the most common malignancy of the female genital tract. MiR-1299 serves as a tumor suppressor, while KCNQ1OT1 acts as an oncogene in multiple malignancies. This research was designed to investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service