Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

373226

Sigma-Aldrich

Hdm2 E3 Ligase Inhibitor II, HLI373

The Hdm2 E3 Ligase Inhibitor II, HLI373, also referenced under CAS 502137-98-6, controls the biological activity of Hdm2 E3 Ligase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Hdm2 E3 Ligase Inhibitor II, HLI373, 5-(3-Dimethylaminopropylamino)-3,10-dimethyl-10H-pyrimido[4,5-b]quinoline-2,4-dione, 2HCl, MDM2 Inhibitor VI, MDM2 E3 Ligase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23N5O2 · 2HCl
CAS Number:
Molecular Weight:
414.33
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

cream

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C1C2=C(NCCCN(C)C)C3=C(C=CC=C3)N(C)C2=NC(N1C)=O.[2HCl]

InChI

1S/C18H23N5O2.2ClH/c1-21(2)11-7-10-19-15-12-8-5-6-9-13(12)22(3)16-14(15)17(24)23(4)18(25)20-16;;/h5-6,8-9,19H,7,10-11H2,1-4H3;2*1H

InChI key

WUEMKAQTAIRDOA-UHFFFAOYSA-N

General description

A cell-permeable pyrimidoquinoline-dione compound that promotes cellular p53 and E3 ubiquitin ligase HDM2/MDM2 built-ups (optimal conc. = 5 µM in human retinal pigment epithelial RPE cells) via direct inhibition of MDM2-mediated p53 ubiquitination and MDM2 autoubiquitination, resulting in an upregulation of p53-dependent transcription activity (2.5-fold of basal in U2OS-based reporter assays; 5 µM 20 h treatment). Shown to induce apoptotic cell death in E1A-transformed, but not non-transformed, RPE cells (72% death; 10 µM 26 h treatment) and be more active toward wild-type p53-expressing than mutant p53-expressing cancer cells (≥67% vs. ≤37% death induction, respectively; 25 µM 30 h treatment).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Kitagaki, J., et al. 2008. Mol. Cancer Ther.7, 2445.
Yang, Y., et al. 2005. Cancer Cell7, 547.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viktoria K Ilic et al.
Biomolecules, 12(5) (2022-05-29)
The proto-oncogene MDM2 is frequently amplified in many human cancers and its overexpression is clinically associated with a poor prognosis. The oncogenic activity of MDM2 is demonstrated by its negative regulation of tumor suppressor p53 and the substrate proteins involved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service