Skip to Content
MilliporeSigma
All Photos(1)

Documents

344220

Sigma-Aldrich

Formyl Peptide Receptor-Like 1 Antagonist, WRW⁴

The Formyl Peptide Receptor-Like 1 Antagonist, WRW⁴ controls the biological activity of Formyl Peptide Receptor-Like 1.

Synonym(s):

Formyl Peptide Receptor-Like 1 Antagonist, WRW⁴, H₂N-WRWWWW-CONH₂, FPRL1 Antagonist, WRW⁴

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H65N15O6
Molecular Weight:
1104.27
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A Trp-rich hexapeptide that acts as a potent and specific antagonist of Formyl Peptide Receptor-Like 1 (FPRL1)-mediated signaling. Shown to bind to FPRL1 and interfere with ligand-receptor interaction {IC50 = 230 nM for FPRL1 Agonist, WKYMVm (Cat. No. 344216)}. Completely blocks ligand-mediated [Ca2+]i increase, ERK activation, and chemotactic migration of FPRL1-expressing RBL-2H3 cells at a concentration of 10 µM. Also inhibits Aβ42 peptide-induced superoxide generation and prevents its internalization in human macrophages.
A Trp-rich hexapeptide that acts as a potent and specific antagonist of Formyl Peptide Receptor-Like 1 (FPRL1)-mediated signaling. Shown to bind to FPRL1 and interfere with the ligand-receptor interaction {IC50 = 230 nM for FPRL1 Agonist, WKYMVm (Cat. No. 344216)}. Completely blocks the ligand activated [Ca2+]i increase, ERK activation and chemotactic migration of FPRL1-expressing RBL-2H3 cells at 10 µM. Further, inhibits Aβ42 peptide-induced superoxide generation and prevents its internalization in human macrophages.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Formyl Peptide Receptor-Like 1 (FPRL1)
Product does not compete with ATP.
Reversible: no
Target IC50: 230 nM for FPRL1 Agonist

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Trp-Arg-Trp-Trp-Trp-Trp-CONH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bae, Y.S., et al. 2004. J. Immunol.173, 607.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service