Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368056

Sigma-Aldrich

Granzyme B Inhibitor IV

The Granzyme B Inhibitor IV controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Granzyme B Inhibitor IV, Ac-IEPD-CHO, Caspase-8 inhibitor III

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$181.00

$181.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
$181.00

About This Item

Empirical Formula (Hill Notation):
C22H34N4O9
Molecular Weight:
498.53
UNSPSC Code:
12352200
NACRES:
NA.77

$181.00


Please contact Customer Service for Availability

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A reversible inhibitor of Granzyme B that also inhibits caspase-8. Granzyme B is a protease involved in the induction of rapid target cell death by cytotoxic lymphocytes.
A reversible inhibitor of granzyme B and caspase-8.

Biochem/physiol Actions

Cell permeable: no
Primary Target
granzyme B
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ile-Glu-Pro-Asp-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Harris, J.L., et al. 1998. J. Biol. Chem. 273, 27364.
Thornberry, N.A., et al. 1997. J. Biol. Chem.272, 17907.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service