Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218745

Sigma-Aldrich

Caspase Inhibitor III

The Caspase Inhibitor III, also referenced under CAS 634911-80-1, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase Inhibitor III, Boc-D-FMK

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$118.00

$118.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
$118.00

About This Item

Empirical Formula (Hill Notation):
C11H18FNO5
CAS Number:
Molecular Weight:
263.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00


Please contact Customer Service for Availability

Quality Level

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FCC(=O)C(NC(=O)OC(C)(C)C)CC(=O)OC

InChI

1S/C11H18FNO5/c1-11(2,3)18-10(16)13-7(8(14)6-12)5-9(15)17-4/h7H,5-6H2,1-4H3,(H,13,16)

InChI key

MXOOUCRHWJYCAL-UHFFFAOYSA-N

General description

A broad-spectrum cell-permeable and irreversible caspase inhibitor.
A cell-permeable, irreversible, broad-spectrum caspase inhibitor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
broad spectrum caspase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Boc-Asp(OMe)-CH₂F*

Reconstitution

Following reconstitution aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 4 months at 4°C or for up to 1 year at -20°C.

Analysis Note

single spot by TLC

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
D’Mello, S.R., et al. 1998. J. Neurochem. 70, 1809.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service