Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36-012-I

Sigma-Aldrich

Anti-nitro-α/α-Synuclein (Tyr39) Antibody, clone nSyn14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

nSyn14, monoclonal

mol wt

calculated mol wt 14.46 kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

mutation (Tyr39)

Gene Information

human ... SCNA(6622)

General description

Alpha-synuclein (UniProt: P37840; also known as Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP) is encoded by the SNCA (also known as NACP; PARK1) gene (Gene ID: 6622) in human. α-synuclein is abundantly expressed in the brain and is found in a classic amyloid fibril form within the intra-neuronal Lewy body deposits of Parkinson′s disease brains. In its monomeric form it participates in synaptic vesicle exocytosis by enhancing vesicle priming, fusion and dilation of exocytotic fusion pores. It acts by increasing local calcium release from microdomains, which is essential for the enhancement of ATP-induced exocytosis. In its multimeric form it acts as a molecular chaperone assisting in the folding of synaptic fusion components called SNAREs at presynaptic membrane. It is also shown to regulate dopamine release and transport. It reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation. Post-translationally it can be phosphorylated, predominantly on serine residues. Phosphorylation of serine 129 is reported to be selective and extensive in synucleinopathy lesions. In addition, increased phosphorylation of Tyr125, nitration of Tyr39, and glycation of α-synuclein have been reported in Parkinsons disease subjects. Nitration of tyrosine residues in α-synuclein is observed in the signature inclusions of Parkinsons disease, dementia with Lewy bodies, the Lewy body variant of Alzheimer s disease, and multiple system atrophy brains. EAAC1 -/- mice display age-dependent loss of dopaminergic neurons in the substantia nigra pars compacta and this neuronal loss is accompanied by increased nitrosylated α-synuclein and microglial activation. Genetic alterations of SNCA gene are reported to result in aberrant polymerization into fibrils, which is associated with several neurodegenerative diseases (synucleinopathies). (Ref.: Miranda, HV., et al. (2017). Sci. Rep. 7; 13713; Berman, AE., et al. (2011). Ann. Neurol. 69(3); 509 520; Giasson, BI., et al. (2000). Science. 290(5493); 985-989).

Specificity

Clone nSyn14 is a mouse monoclonal antibody that detects a/b synuclein nitrated at tyrosine 39.

Immunogen

Full-length recombinant human α-synuclein nitrated at tyrosine 39.

Application

Anti-nitro-α/β-Synuclein (Tyr39), clone nSyn14, Cat. No. 36-012-I, is a mouse monoclonal antibody that detects α/β-Synuclein nitrated on tyrosine 39 and is tested for use in ELISA, Immunohistochemistry, and Western Blotting.
Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1.

Tested Applications

ELISA Analysis: A representative lot detected nitro- α/β-Synuclein (Tyr39) in ELISA applications (Giasson, B.I., et al. (2000). Science. 290(5493):985-9).
Immunohistochemistry Analysis: A representative lot detected nitro- α/β-Synuclein (Tyr39) in Immunohistochemistry applications (Maki, R.A., et al. (2019). Free Radic Biol Med. 141:115-140; Giasson, B.I., et al. (2000). Science. 290(5493):985-9).
Western Blotting Analysis: A 1:500 dilution from a representative lot detected nitro- α/β-Synuclein in Various a-synuclein protein samples (chemically nitrated or untreated) (Courtesy of Sarah Wright, Nitrome Biosciences, Inc., Brisbane, CA USA).
ELISA Analysis: Various dilutions of this antibody detected nitro- α/β-Synuclein in various a-synuclein protein samples (chemically nitrated or untreated) (Courtesy of Sarah Wright, Nitrome Biosciences, Inc., Brisbane, CA USA). .
Western Blotting Analysis: A representative lot detected nitro- α/β-Synuclein (Tyr39) in Western Blotting applications (Miranda, H.V., et al. (2017). Sci Rep. 7(1):13713; Fernandez, E., et al. (2014). Antioxid Redox Signal. 21(15):2143-8; Hung, L.W., et al. (2012). J Exp Med. 209(4):837-54; Giasson, B.I., et al. (2000). Science. 290(5493):985-9).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service