Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCP4MG64KPX20BK

Millipore

MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel IV Premix Space Saver (Bulk) Packaging

Sign Into View Organizational & Contract Pricing

Select a Size

96 -WELL PLATE
$6,150.00

$6,150.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 -WELL PLATE
$6,150.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

$6,150.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Inflammation and immune cell response are key to maintaining homeostasis. The more we understand mechanisms of disease, the more we discover the role that inflammation plays in the etiology of disease: certainly immunological diseases, such as allergic reactions, asthma, infectious disease and an ever-growing number of conditions under the autoimmune umbrella, but also neurodegenerative disease, metabolic and cardiovascular disease and even cancer. The ability to measure multiple inflammation biomarkers simultaneously using MILLIPLEX® MAP panels, based on the Luminex xMAP technology, to establish disease profiles that can be used for screening, diagnosis and response to treatment is critical for understanding the pathogenesis of these diseases. MILLIPLEX® MAP Cytokine/Chemokine Magnetic Bead Panel IV Space Saver/Bulk Packaging Premix is a 20-plex premixed kit to be used for the simultaneous quantification of all of the following analytes in serum, plasma or culture supernatant samples: BAFF/Blys, BRAK/CSCL14, CCL28, CXCL16, HMGB1, IFNβ, IL-14, IL-19, IL-24, IL-28B, IL-32α, IL-34, IL-35, IL-36β, IL-37, IL-38, MIP-4/PARC/CCL18, MPIF-1/CL23 and YKL40/CHI3L1MILLIPLEX® MAP offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust. In addition to the assay characteristics listed in the protocol, other performance criteria evaluated during the validation process include: cross-reactivity, dilution linearity, kit stability, and sample behavior (e.g. detectability and stability). Each panel and kit meets stringent manufacturing criteria to ensure batch-to-batch reproducibility.Each MILLIPLEX® MAP panel and kit includes:Quality controls (QCs) provided to qualify assay performanceComparison of standard (calibrator) and QC lots to a reference lot to ensure lot-to-lot consistencyOptimized serum matrix to mimic native analyte environmentDetection antibody cocktails designed to yield consistent analyte profiles within panelMILLIPLEX® MAP Human Cytokine/Chemokine Magnetic Bead Panel IV is part of the most versatile system available for cytokine research. From our single to multiplex biomarker solutions, we partner with you to design, develop, analytically validate and build the most comprehensive library available for protein detection and quantitation.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The Human Inflammation / Immunology Bead-Based Multiplex Cytokine/Chemokine Bulk Packaging Premix Assay kit IV, using the Luminex® xMAP technology, enables the simultaneous analysis of 20 cytokine and chemokine biomarkers in human serum, plasma and cell culture samples.This is an overnight or two-hour incubation assay; however, for best results an overnight incubation is recommended.A maximum of 25 μL per well of 1:4 diluted serum or plasma can be used. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay. Please note that HMGB1 can be measured in serum and plasma samples only.Analytes available:BAFF/Blys;BRAK/CXCL14;CCL28;CXCL16;HCC-4/CCL16;HMGB1;IFNβ;IL-14;IL-19;IL-24;IL-28B/IFNλ3;IL-32α;IL-34;IL-35;IL-36β/IL-1F8;IL-37/IL-1F7;IL-38/IL-1F10;MIP-4;MPIF-1/CCL23;YKL40

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service