Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345832

Sigma-Aldrich

Galectin-3C, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$1,010.00

$1,010.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$1,010.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$1,010.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤1 EU/μg protein Endotoxin

solubility

sterile PBS: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human galectin-3 expressed in E. coli and subsequently cleaved with Clostridium histolyticum collagenase type VII to yield galectin-3C. The resulting galectin-3C is purified by affinity chromatography. Galectin-3C is a truncated form of galectin-3 that contains the carbohydrate binding domain but lacks the 107 amino acids at the N-terminus of galectin-3. It is suggested to play a key role in autoimmune diseases, allergic reactions, inflammation, tumor cell metastasis, atherosclerosis, and diabetes.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from deionized water, 2µg lactose/µg lectin.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions (in the presence of a molar excess of lactose) are stable for up to 4 months at -20°C. For use in carbohydrate binding assays the solution should be dialyzed to remove the lactose stabilizer.

Other Notes

Hoyer K.K., et al. 2004. Am. J. Pathol.164, 893.
John C.M., et al. 2003. Clin. Cancer Res.9, 2374.
Iurisci, I., et al. 2000. Clin Cancer Res6, 1389.
Van Den Brule, F. A., et al. 2000. Int. J. Cancer89, 361.
Inohara, H. 1999. Cancer85, 2475.
Nakamura, M., et al. 1999. Int. J. Oncol.15, 143.
Idikio, H., et al. 1998. Int. J. Oncol.12, 1287.
Ochieng, J., et al. 1998. Biochim. Biophys. Acta.1379 97.
Castronovo, V., et al. 1996. J. Pathol.179, 43.
Liu, F.T., et al. 1996. Biochemistry35 6073.
Schoeppner, H. L., et al. 1995. Cancer75, 2818.
Massa, S.M., et al. 1993. Biochemistry32, 260.
Oda, Y., et al. 1991. Gene99, 279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service