Skip to Content
MilliporeSigma
All Photos(1)

Documents

70856

Millipore

RNase A Solution

Convenient solution for selective degradation of RNA

Synonym(s):

RNase A Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

10 mg/mL

technique(s)

DNA purification: suitable

shipped in

ambient

storage temp.

10-30°C

General description

RNase A Solution is a convenient alternative to powdered RNase A. It is a highly purified preparation of bovine pancreatic ribonuclease A suitable for use in selective removal of RNA. It has been pretreated to remove DNase I and is suitable to remove RNA contamination during plasmid and genomic DNA purification procedures. Supplied at a concentration of 10 mg/ml in 10 mM Tris-HCl, 1 mM EDTA, 50% glycerol, pH 7.5.

Application

RNase A Solution has been used:
  • in cell cycle profiling of embryonic mouse neuroectodermal cells (NE-4C)
  • in the digestion of RNA for extraction of DNA from human osteosarcoma cell lines for chromatin immunoprecipitation (ChIP) assays
  • to treat control adult and embryonic thymi cells during staining
  • to remove RNA from affibody–mRNA fusion clones before clone screening

Biochem/physiol Actions

RNaseA is an endoribonuclease that catalyzes the degradation of phosphodiester linkages with a pyrimidine base at the 3′ -position on single-stranded RNA via, transphosphorylation and hydrolysis mechanism.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurotrophic properties and the de novo peptide sequencing of edible bird's nest extracts
Yew M Y, et al.
Food Bioscience, 32, 100466-100466 (2019)
Myc controls a distinct transcriptional program in fetal thymic epithelial cells that determines thymus growth
Jennifer E C, et al.
Nature Communications (2019)
Jiajun Zhang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(11), 2681-2692 (2020-01-16)
Malignancy of cancer cells depends on the active transcription of tumor-associated genes. Recently, unique clusters of transcriptional enhancers, termed super-enhancers, have been reported to drive the expression of genes that define cell identity. In this study, we characterized specific super-enhancer-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service