Skip to Content
MilliporeSigma
All Photos(1)

Documents

218682

Sigma-Aldrich

Casein, Bovine Milk, Carbohydrate and Fatty Acid Free

Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, is a high molecular weight phosphoprotein. Useful as a substrate for various proteases. Supplied as Hammarsten grade material.

Synonym(s):

Casein, Bovine Milk, Carbohydrate and Fatty Acid Free

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

Assay

≥95% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤0.1% Fatty acids
≤0.1% free acid
≤0.5% glucose
≤0.5% lactose

color

white to off-white

solubility

dilute base: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

General description

High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases. Supplied as Hammarsten grade material.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C. After one week, it is recommended that fresh solution be prepared.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service