Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218680

Sigma-Aldrich

Casein from bovine milk

protease substrate, solid

Synonym(s):

Casein, Bovine Milk

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Casein, Bovine Milk, Casein, Bovine Milk, CAS 9000-71-9, is a high molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases.

Quality Level

description

Merck USA index - 14, 1883

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

beige

solubility

dilute base: 1 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

General description

Casein is a major protein present in milk and constitutes up to 80% of total milk protein.[1] It is a calcium phosphate-binding phosphoprotein present in mammalian milk. Casein in bovine milk is made of four types of peptides as αS1, αS2, β, and κ.[2]

Biochem/physiol Actions

Casein has several uses as in adhesives, coatings and packaging material in the paper and food industries.[1]

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun Ghosh et al.
Biomacromolecules, 10(7), 1681-1688 (2009-05-14)
The development of advanced materials from biorenewable protein biopolymers requires the generation of more exogenous bonds to maintain the microstructure and durability in the final products. Casein is the main protein of milk, representing about 80% of the total protein.
Tomasz Konrad Głąb et al.
Topics in current chemistry (Cham), 375(4), 71-71 (2017-07-18)
Casein is the collective name for a family of milk proteins. In bovine milk, casein comprises four peptides: αS1, αS2, β, and κ, differing in their amino acid, phosphorus and carbohydrate content but similar in their amphiphilic character. Hydrophilic and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service