Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208291

Millipore

Calcium chloride dihydrate

for molecular biology

Synonym(s):

CaCl₂

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$59.60

$59.60


Available to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$59.60

About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25
grade:
Molecular Biology
for molecular biology
form:
solid
solubility:
water: 150 mg/mL

$59.60


Available to ship onMarch 31, 2025Details


Request a Bulk Order

grade

Molecular Biology
for molecular biology

Quality Level

description

Merck USA index - 14, 1659

Assay

≥99% (complexometric)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

mp

176 °C (dec.) (lit.)

solubility

water: 150 mg/mL

cation traces

heavy metals (as Pb): ≤0.0005%

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

General description

Has an optimal pH of 5.0-8.0.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service