Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93205

Supelco

Potassium phosphate monobasic

for analysis EMPARTA® ACS

Synonym(s):

Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
12352302
grade:
ACS reagent
Pricing and availability is not currently available.

grade

ACS reagent

product line

EMPARTA®

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93205.0521: 500G, Minimum order quantity: 20
1.93205.5021: 5 kg, Minimum order quantity: 1
1.93205.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service