Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

178670

Millipore

D(–)Arabinose

An uncommon pentose that can be utilized as a carbon source by bacteria.

Synonym(s):

D(–)Arabinose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352201

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2

InChI key

SRBFZHDQGSBBOR-UHFFFAOYSA-N

General description

An uncommon pentose that can be utilized as a carbon source by bacteria.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, filter-sterilize and store at room temperature (20°C). Stock solutions are for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service