Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

178462-M

Sigma-Aldrich

Apolipoprotein C-II, Human Plasma, Very Low-Density Lipoprotein

Native apolipoprotein C-II from human plasma. Found primarily in VLDL and chylomicrons. Functions as a cofactor for lipoprotein lipase.

Synonym(s):

Apo C-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

storage temp.

−20°C

General description

Native apolipoprotein C-II from human plasma. Found primarily in VLDL and chylomicrons. Functions as a cofactor for lipoprotein lipase. Elevated serum levels of Apo C-II are common in type I, III, IV, and V hyperlipoproteinemia. Low serum concentrations of Apo C-II are indicative of nephrotic syndrome (Tangier’s Disease).

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 10 mM NH₄HCO₃, pH 7.4.

Preparation Note

Please refer to vial label for lot-specific volume prior to lyophilization.

Other Notes

Brunzell, J.D. 1989. in The Metabolic Basis of Inherited Disease,6th Edition (Scriver, G.R., ed.) p. 1165, McGraw Hill, New York.
Le, N.A., et al. 1988. J. Lipid Res. 29, 669.
Jackson, R.L., and Holdsworth, G. 1986. Methods Enzymol.128, Chapter 14.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service