Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152224

Sigma-Aldrich

AEC

This AEC is validated for use in Immunoblotting and Immunohistochemistry.

Synonym(s):

AEC, 3-Amino-9-ethylcarbazole, 3AC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
41105333

Quality Level

form

solution

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-brown to reddish

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H14N2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2,15H2,1H3

InChI key

OXEUETBFKVCRNP-UHFFFAOYSA-N

General description

Precipitable peroxidase substrate for immunoblotting and immunohistochemical staining techniques. Produces a red end product that is soluble in alcohol. When used in immunohistochemistry, aqueous mounting media, and counterstains must be used. Directions included for use in each application.

Application

Immunoblotting (see comments)

Immunohistochemistry (see comments)

Warning

Toxicity: Toxic (F)

Reconstitution

Discard if a red or brown precipitate is evident.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Okano, K., et al. 1992. Rinsho Byori 40, 93.
Krenacs, T., et al. 1990. Histochem. J. 22, 530.
Stable at 4°C and 18-26°C. Functionality determined by performing an immunohistologic stain using the final product.



Suggested procedure for use of AEC in immunohistochemistry and immunoblotting:



1. Complete all required incubations with antibodies and HRP-labeled probes, and all necessary wash steps.

2.Prepare 1X AEC Substrate Buffer: Add 9 parts distilled water to 1 part 10X AEC Substrate Buffer, Cat. No. 152226 (0.5 M acetate buffer containing 88 mM H2O2). Mix well. The final pH of this solution will be ~5.0.

3. Prepare 1X AEC Substrate Solution: Add 1 part 50X AEC Concentrate to 50 parts 1X AEC Substrate Buffer (prepared in Step 2). Mix well.

4. Following the final wash, completely cover tissue sections with 1X AEC Substrate Solution (prepared in Step 3) and incubate 10-30 min at room temperature. The same conditions apply for immunoblotting. Note: Variables associated with assay conditions will dictate the proper reaction times.

5. After reaction is complete, wash tissue sections or blotting membranes thoroughly in distilled water.

6. If tissue sections require a counterstain, Mayer′s Hematoxylin followed by blueing in alkaline tap water or a 0.5% aqueous methyl green solution is recommended.

7. Mount tissue sections in a water-based mounting medium. Air-dry blots and store protected from light.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service