Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46592

Millipore

Phosphate Buffer Solution

bottle of, ready-to-use, bottle volume 1000 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw 3 loci with septum
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable

bottle capacity

1000 mL

bottle volume

1000 mL , filling volume

pH

7.4± 0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Phosphate buffer is designed to cater as a sample diluent and a solution for performing serial dilution. It prevents pH-associated variations while carrying out microbial experiments.

Application

Phosphate buffer is used for the preparation of dilutions, blanks for the examination of waters, dairy products, foods, and other test samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service