Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46121

Millipore

ReadyTube® Yeast Extract Agar

For the enumeration of culturable microorganism in water samples in accordance with ISO 6222, pkg of (18 ml in 25 ml tube with blue cap (20 or 100 tubes per box))

Synonym(s):

YE Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

Agency

ISO 6222

Quality Level

product line

ReadyTube®

feature

ready-to-use

packaging

pkg of (18 ml in 25 ml tube with blue cap (20 or 100 tubes per box))

pH

7.2 ( in H2O)

application(s)

bioburden testing
food and beverages
microbiology
water monitoring

storage temp.

2-25°C

suitability

nonselective for

General description

Yeast extract agar is a medium rich in nutrients that permits the recovery of a large spectrum of bacteria, yeast, and molds. ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® Yeast extract agar is a highly nutritive medium recommended for the plate count of microorganisms in water. Water contains large number of microorganisms, the combination of a nutrient-rich culture medium and incubation temperatures of +22 °C and +36 °C allows the detection of many of these organisms. The media contains Yeast extract agar formulation that is in accordance with ISO 6222. The peptone in the media provides nitrogenous compounds, vitamin B complex, and other growth nutrients.

Application

ReadyTube® Yeast extract agar is a medium rich in nutrients, which permits the recovery of a wide spectrum of bacteria, yeast, and molds from water samples. The media finds application in the food and beverage industry

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service