Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10878

Millipore

GranuCult® prime Count agar sugar free

according to ISO 13559 I IDF 153:2022, for heterotrophic bacteria

Synonym(s):

Bacterial count agar sugar-free, Bacterial count agar sugar-free, Count agar sugar free, Colony Count Agar sugar-free, Count agar sugar free, Sugar Free Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$311.00

$311.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
$311.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

$311.00


Please contact Customer Service for Availability

Request more information

Agency

according to ISO 13559 I IDF 153:2022

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

dry media (Keep tightly closed.)

technique(s)

microbiological culture: suitable

color

beige

pH

7.4-7.6 (37 °C, 35 g/L in H2O, after autoclaving)

solubility

35 g/L

bulk density

700 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

heterotrophic bacteria
infectious bacteria (in dairy)

General description

GranuCult® prime Count agar sugar-free ISO 13559 I IDF 153:2022, is used to determine the count of infective microorganisms in butter and other dairy products. Infective microorganisms are defined as organisms that are not directly involved in the microbiological production of a dairy product or that do not belong to its specific flora. This culture medium does not contain any fermentable carbohydrates and has little nutrient value, so these microorganisms can be cultivated selectively.

Application

GranuCult® prime Count agar sugar free ISO 13559 I IDF 153:2022, is suitable for determining the count of infective microorganisms in butter and other dairy products.

Features and Benefits

  • GranuCult® offers superior granulated culture media.
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions.
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear to slightly opalescent
Appearance (colour): yellowish
pH-value (25 °C): 7.3 - 7.7
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 70 %
Recovery on test medium (Staphylococcus aureus ATCC 25923 (WDCM 00034)): ≥ 70 %
Recovery on test medium (Enterococcus faecalis ATCC 11700): ≥ 70 %
Recovery on test medium (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): ≥ 70 %
Recovery on test medium (Bacillus cereus ATCC 11778 (WDCM 00001)): ≥ 70 %
Recovery on test medium (Candida albicans ATCC 10231 (WDCM 00054)): ≥ 70 %
Incubation: 48 hrs.; 35 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service