Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44210

Millipore

Table-top Tripod

for use with RCS® High Flow Touch

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$139.00

$139.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$139.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

$139.00


Please contact Customer Service for Availability

material

metal stand

Agency

non-compendial

feature

adjustable

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

shipped in

ambient

storage temp.

room temp

General description

Table-top Tripod is an additional accessory for use with RCS® microbial air sampler. The RCS® portable air sampler is used for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The Table-top Tripod is used for horizontal operation of the Air Sampler on very smooth surfaces to minimize vibrations.

Application

Table-top Tripod is an additional accessory with the RCS® microbial air sampler used for sampling on very smooth surfaces to minimize vibrations. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service