Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09326

Millipore

Tripod

For use with portable MAS-100 family, length 88-250 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

metal stand

Agency

non-compendial

product line

MAS-100®

feature

adjustable

packaging

pkg of 1 kit

L

88-250 cm

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with Portable Air Samplers

storage temp.

2-30°C

General description

Tripod for MAS-100, length 88-250 cm couple with all portable MAS-100® air samplers either directly via screwing or in combination with the quick-change adapter for air sampling up to 250 cm height. Whether as a mobile and stable work surface for microbial air sampling in pharmaceutical cleanrooms or as an extension to reach the ceiling air outlets during hygiene inspections, the height is adjustable and three- legged tripod can reach any height ranging from 88-250 cm effortlessly. For safety information of selected height please refer to the individual air sampler operation manuals. The tripod can easily be combined with the quick-change adapter for all portable MAS-100® viable air samplers to avoid screwing for each single connection or with a specific adapter for the MAS-100 Eco® This distributes the load stably on the three supports and the microbial air sampling enabling air sampling at airy height easier than ever.

Application

Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as in Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service