Skip to Content
MilliporeSigma

1.37039

SAFC

Potassium dihydrogen phosphate

crystalline, EMPROVE® EXPERT, Ph. Eur., BP, JPC, NF

Pharma Manufacturing

Synonym(s):

Potassium phosphate monobasic, mono-Potassium orthophosphate, Potassium biphosphate, Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-913-4

Agency

BP
JPC
NF
Ph. Eur.

Quality Level

product line

EMPROVE® EXPERT

Assay

≥99.0% dry basis (alkalimetric)
98.0-100.5% (alkalimetric)

form

crystalline
solid

feature

RNase and DNase free

pH

4.2-4.6 (20 °C, 20 g/L in H2O)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium dihydrogen phosphate is typically used as a phosphate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.
Due to its low microbial and endotoxin limits, Potassium dihydrogen phosphate Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service