Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.32370

Millipore

Anaerocult C mini

Incubation bags for creating a low-oxygen, high-CO2 atmosphere, suitable for microbiology

Synonym(s):

Microaerophilic growth, Microaerophilic incubation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

Quality Level

form

solid

feature

Produces a low-oxygen and high-CO2 atmosphere.

manufacturer/tradename

Anaerocult

packaging

pkg of 25 Anaerocult C mini and 25 special incubation bags

storage condition

(Store dry and tightly closed)

color

white

application(s)

microbiology

compatibility

ANAEROTEST

storage temp.

15-25°C

General description

Anaerocult C Mini contains chemicals that bind a precisely determined amount of oxygen in the special incubation bag and release a defined quantity of CO2, producing an oxygen-depleted, CO2-enriched atmosphere. This creates the right atmospheric conditions for the microaerophilic or capnophilic microorganisms to grow.The Anaerocult C Mini special incubation bags, which are included with the product, can hold two Petri dishes. These can be two different culture media plates inoculated with one sample or two plates with the same medium inoculated with two different samples. The Anaerocult C Mini sachet is moistened with 3 ml of water and placed in the special incubation bag with the Petri dishes.

Application

Anaerocult C mini is a gas generator system for the incubation of one or two Petri dishes in a low-oxygen, high-CO2 atmosphere for the growth of anaerobic microorganisms.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service