Skip to Content
MilliporeSigma
All Photos(4)

Documents

1.32382

Millipore

Anaerocult P

For creating an anaerobic environment in a single Petri dish for the cultivation of obligate and facultative anaerobes, suitable for microbiology

Synonym(s):

Anaerobic growth, Anaerobic incubation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

Quality Level

form

solid

feature

Creates an oxygen free (anaerobic) environment and a CO2 atmosphere in a single petri dish.

manufacturer/tradename

Anaerocult

packaging

pkg of 25 Anaerocult P and 25 special incubation bags

storage condition

(Store dry and tightly closed.

Keep in a well-ventilated place.)

suitability

suitable for (Anaerobic bacteria)
anaerobic bacteria

application(s)

food and beverages
microbiology

compatibility

ANAEROTEST

storage temp.

15-25°C

General description

Anaerocult P device is used for anaerobic incubation of individual Petri dishes. Anaerotest strip (1.32371.0001) attached to the lid of the Petri dish as well as Anaerocult P sachet is moistened with water and placed in a special incubation bag, which is subsequently sealed. It contains components that chemically bind oxygen quickly and completely, creating an oxygen-free environment and a CO2-containing atmosphere. Each Petri dish can be incubated immediately after receiving the sample. This sachet is ideal for both urgent cases and the cultivation of extremely oxygen-sensitive microorganisms. Incubation of individual Petri dishes permits each one to be cultivated separately over time and at the temperature required. It also prevents interactions between the microorganisms on different plates. The transparent incubation bag allows anaerobic growth in the closed anaerobic system to be evaluated visually at any given time.

Application

AnaerocultP is a microbiological reagent for generating an anaerobic atmosphere in a single Petri dish to permit the cultivation of obligate and facultative anaerobes.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service