Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1218

Sigma-Aldrich

Buffer, Phosphate pH 6.8

HARLECO® Used to dilute hematology and histology stains

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703

Quality Level

form

liquid

color

colorless

pH

6.8 ( in H2O)

mp

-0.1 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

General description

HARLECO® Buffer, Phosphate pH 6.8
1218-75 1L Poly Bottle

Application

Buffer used in Wright and Wright-Giemsa staining protocols.
HARLECO® Used to dilute hematology and histology stains

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service