Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.20593

Millipore

modified Tryptic Soy Broth- Dehydrated Culture Media

Novobiocin, for enterovirulent E. coli (EEC)

Synonym(s):

mTSB broth Selective Supplement, modified Tryptic Soy Broth Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

Agency

USDA-FSIS

Quality Level

form

solid

composition

Novobiocin

manufacturer/tradename

Millipore

pH range

7.0 - 7.4 (25 °C)

application(s)

pathogen testing

storage temp.

2-8°C

suitability

enterovirulent E. coli (EEC) (Antibiotics for 1.40011 mTSB culture medium)

Looking for similar products? Visit Product Comparison Guide

General description

mTSB broth Selective Supplement contains Novobiocin sodium salt (8 mg) in lyophilized form. The supplement is suitable for suppressing the growth of accompanying Gram-positive bacteria during culturing of E. coli (O157:H7).

Application

mTSB broth Selective Supplement is to be used with GranuCult® mTSB (modified Tryptic Soy Broth) acc. USDA-FSIS (1.40011) to suppress the growth of accompanying flora of Gram-positive bacteria during culturing Escherichia coli O157:H7.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service