Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.18408

Millipore

Tryptic Soy agar with Neutralizers

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

Agency

EP
USP

form

solid

application(s)

microbiology

storage temp.

20-25°C

General description

Contact plates Envirocheck

Analysis Note

Appearance:clear, yellowish
pH-value:7.1 - 7.5
Irradiation dose:9 - 20 kGy
Sterility:no growth
Filling volume (ml):17 ml
Growth Promotion Test according to EP, USP and JP.
Inoculum on reference medium
- Staphylococcus aureus ATCC 6538 (WDCM 00032):
10 - 100
- Escherichia coli ATCC 8739 (WDCM 00012):10 - 100
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026):10 - 100
- Bacillus subtilis ATCC 6633 (WDCM 00003):10 - 100
- Candida albicans ATCC 10231 (WDCM 00054):10 - 100
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053):10 - 100
Recovery on test medium
- Staphylococcus aureus ATCC 6538 (WDCM 00032):
50 - 200 %
- Escherichia coli ATCC 8739 (WDCM 00012):50 - 200 %
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026):50 - 200 %
- Bacillus subtilis ATCC 6633 (WDCM 00003):50 - 200 %
- Candida albicans ATCC 10231 (WDCM 00054):50 - 200 %
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053):50 - 200 %
Incubation: 24 hrs.; 30-35 °C; aerobic; Candida 2 days; 20-25 °C; Aspergillus 3 days; 20-25 °C. Reference medium: Tryptic Soy Agar
The information above is current at this time of publication and is subject to change without notice (except for customers holding a change control agreement with our company). The information/format can be altered at any time and in any way if internal company matters or Standard Regulations do say so.

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service